Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TNFSF11 / RANKL / CD254 Protein, Fc Tag, Avi Tag (Avitag™);HEK293;200UG;RAL,TNFSF11,RANKL,CD254,TRANCE,OPGL,ODF
Catalog No. 501657552
Encompass_Preferred
Recombinant Protein;Biotinylated Human TNFSF11 / RANKL / CD254 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human TNFSF11, Fc Tag, Avi Tag (RAL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 49.2 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, Fc Tag, Avi Tag (Cat. No. RAL-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-7552 Supplier ACROBiosystems Supplier No. RALH82F9200UG
Recombinant Protein;Biotinylated Human TNFSF11 / RANKL / CD254 Protein, Fc Tag, Avi Tag (Avitag™);200UG;Biotinylated Human TNFSF11, Fc Tag, Avi Tag (RAL-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries an Avi tag (Avitag™) at the N-terminus, followed by a human IgG1 Fc tag. The protein has a calculated MW of 49.2 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Biotinylated Human TNFSF11, Fc Tag, Avi Tag (Cat. No. RAL-H82F9) with a linear range of 0.4-13 ng/mL (QC tested).