Learn More
ACROBiosystems Recombinant Protein;Human TNFSF11 / RANKL / CD254 Protein, Fc tag;HEK293;1MG;RAL,TNFSF11,RANKL,CD254,TRANCE,OPGL,ODF
Supplier:  ACROBiosystems RALH52651MG
Encompass_Preferred
Recombinant Protein;Human TNFSF11 / RANKL / CD254 Protein, Fc tag;1MG;Human TNFSF11, Fc tag (RAL-H5265) is expressed from human 293 cells (HEK293). It contains AA Gly 64 - Asp 245 (Accession # AAC51762.1).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 46.9 kDa. The protein migrates as 60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human RANK, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. RAK-H5251) at 0.5 μg/mL (100 μL/well) can bind Human TNFSF11, Fc tag (Cat. No. RAL-H5265) with a linear range of 0.1-3 ng/mL (QC tested).;Immobilized Human OPG, His Tag (Cat. No. TNB-H5220) at 10 μg/mL (100 μL/well) can bind Human TNFSF11, Fc tag (Cat. No. RAL-H5265) with a linear range of 0.3-2 ng/mL (Routinely tested).
Catalog No. 50-165-7882