Learn More
Aviva Systems Biology Corp TNFSF12 Antibody - N-terminal region (ARP36558_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36558_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFSF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNFSF12. Alternate names: APO3L, DR3LG, MGC129581, MGC20669, TWEAKApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56188