Learn More
Aviva Systems Biology Corp TNFSF13 Antibody - middle region (ARP60977_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60977_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFSF13. It was validated on Western Blot. . Alternate names: APRIL, CD256, TALL2, TRDL-1, UNQ383/PRO715, ligand, ZTNF2, TALL-2ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-219