Learn More
Aviva Systems Biology Corp TNFSF14 Antibody - middle region (AVARP02041_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP02041_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFSF14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNFSF14. Alternate names: CD258, HVEML, LIGHT, LTg, TR2ApplicationWB. Species reactivity: Dog
Catalog No. 50-014-62487