Learn More
Aviva Systems Biology Corp TNFSF4 Antibody - middle region (ARP59092_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59092_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNFSF4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TNFSF4. Alternate names: CD134L, CD252, GP34, OX-40L, OX4OL, TXGP1ApplicationWB. Species reactivity: Human
Catalog No. 50-286-574