Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse TNFSF4 (OX40L) ELISA Kit

Catalog No. EMTNFSF4X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMTNFSF4X5 5 x 96 Tests
EMTNFSF4 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EMTNFSF4X5 Supplier Invitrogen™ Supplier No. EMTNFSF4X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

The Mouse TNFSF4 (OX40L) ELISA quantitates Ms TNFSF4 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms TNFSF4. Principle of the method The Mouse TNFSF4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The tumor necrosis factor superfamily member TNFSF4 is a type II membrane bound, non-covalently linked homotrimeric protein. It is expressed on antigen presenting cells, such as dendritic cells and activated B-cells, and also on other cells such as vascular endothelial cells, mast cells, and natural killer cells. TNFSF4 signals specifically through the TNFRSF4 receptor, is expressed predominantly on CD4+T cells but also on certain activated CD8+T cells. TNFRSF4/TNFSF4 functions as a costimulatory signal, which is required for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival, and increases expression of RANTES, IL-2, IL-3, and IFNgamma. TNFRSF4/TNFSF4 signaling plays an important role in immunotolerance.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P43488
Assay Range 30-8000 pg/mL
Assay Sensitivity 30 pg/mL
Calibration Range 30 pg/mL to 8000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA pair antibody detects mouse TNFSF4/OX40 Ligand and on cross reactivity with rmBAFF, rmEDA, rmGITR Ligand, rmFas Ligand, rmLIGHT, rmTWEAK, rhAPRIL, or rhTNFSF4.
Sufficient For 96 assays
Target Species Mouse
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Ath1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1lAth1; Ath-1; atherosclerosis 1; CD134 ligand; CD134L; CD252; glycoprotein Gp34; GP34; OX40 antigen ligand; OX40 ligand; OX40L; OX-40L; OX4OL; TAX transcriptionally-activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 (34kD); tax-transcriptionally activated glycoprotein 1 ligand; TNFSF4; TNLG2B; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor (ligand) superfamily, member 4; tumor necrosis factor ligand 2B; tumor necrosis factor ligand superfamily member 4; tumor necrosis factor superfamily member 4; TXGP1; Txgp1l; Ath-1; Ath1; CD134L; OX-40L; Ox40l; TXGP1; Tnlg2b; Txgp1l; gp34
Gene ID (Entrez) 22164
Gene Symbol Tnfsf4
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases OX40 ligand, OX40L
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym Ath-1, Ath1, CD134L, OX-40 L, O x 40 L, TXGP1, Txgp1l, gp34
Target OX40L (CD252)
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.