Learn More
Cayman Chemical TNFSF8CD30 LIgnd EXrAClu 200ug
SDP
Catalog No. 502942395
Encompass_Preferred
Source: Active recombinant N-terminal human IgG1 Fc-tagged CD30L expressed in HEK293 cells • Amino acids: 63-234 • MW: 47.8 kDa
Catalog No. 50-294-2395 Supplier Cayman Chemical Supplier No. 40213200 UG
Source: Active recombinant N-terminal human IgG1 Fc-tagged CD30L expressed in HEK293 cells • Amino acids: 63-234 • MW: 47.8 kDa