Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TNG-0746132 | 2821749-57-7 | C26H22F3N7O | 5 MG
SDP

Catalog No. 5000372624
Encompass_Preferred

TNG-0746132 is a compound that can be used for the synthesis of other compounds with anticancer activity. It is intended for research use only and is not for direct sale to patients.

  • High purity (99.48%)
  • Soluble in DMSO at 33.33 mg/mL
  • Long-term storage stability as powder: 3 years at -20°C, 2 years at 4°C
  • Storage stability in solvent: 6 months at -80°C, 1 month at -20°C

Catalog No. 50-003-72624 Supplier Medchemexpress LLC Supplier No. HY1533665MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TNG-0746132 is a compound that can be used for the synthesis of other compounds with anticancer activity. It is intended for research use only and is not for direct sale to patients.

  • High purity (99.48%)
  • Soluble in DMSO at 33.33 mg/mL
  • Long-term storage stability as powder: 3 years at -20°C, 2 years at 4°C
  • Storage stability in solvent: 6 months at -80°C, 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.