Learn More
Electron Microscopy Sciences EMS Tungsten Carbide Mayo Scissors with Curved Blades 5-1/2"
SDP
Catalog No. 501931439
Encompass_Preferred
These Mayo scissors are made from Tungsten Carbide and have blunt rounded tips. The unique tungsten carbide cutting edge provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved
OAL: 5-1/2"
This item is not returnable. View return policy
Catalog No. 50-193-1439 Supplier Electron Microscopy Sciences Supplier No. 7299632
These Mayo scissors are made from Tungsten Carbide and have blunt rounded tips. The unique tungsten carbide cutting edge provides effortless, yet accurate, cuts through the toughest of surgical or autopsy tissue. Scissor shanks are tempered to the ideal hardness to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue during complex dissection cases.
Blade: Curved
OAL: 5-1/2"