Learn More
TARGETMOL CHEMICALS INC TNIK-IN-2 5MG
SDP
Catalog No. 502579000
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TNIK-IN-2 is an inhibitor Traf2- and Nck-interacting protein kinase (TNIK IC50 = 1.33 uM)) which is a downstream signal protein of the Wnt/(beta)-catenin pathway. purity: 98%
Catalog No. 50-257-9000 Supplier TARGETMOL CHEMICALS INC Supplier No. T95575MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TNIK-IN-2 is an inhibitor Traf2- and Nck-interacting protein kinase (TNIK IC50 = 1.33 uM)) which is a downstream signal protein of the Wnt/(beta)-catenin pathway. purity: 98%