Learn More
TARGETMOL CHEMICALS INC TNIK-IN-5 5MG
SDP
Catalog No. 502578955
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TNIK-IN-5 is an potent TNIK inhibitor (IC50= 0.05 uM). TNIK-IN-5 potently inhibits Wnt signaling in intact cells. TNIK-IN-5 exhibits excellent in vitro anti-colorectal cancer activity. purity: 99%
Catalog No. 50-257-8955 Supplier TARGETMOL CHEMICALS INC Supplier No. T98105MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TNIK-IN-5 is an potent TNIK inhibitor (IC50= 0.05 uM). TNIK-IN-5 potently inhibits Wnt signaling in intact cells. TNIK-IN-5 exhibits excellent in vitro anti-colorectal cancer activity. purity: 99%