Learn More
Aviva Systems Biology Corp TNIP1 Antibody - C-terminal region (ARP61714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNIP1. It was validated on Western Blot. . Alternate names: ABIN-1, KIAA0113, NAF1, VAN, nip40-1ApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-287-445