Learn More
Aviva Systems Biology Corp TNIP2 Antibody - N-terminal region (ARP64997_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64997_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNIP2. It was validated on Western Blot. . Alternate names: ABIN2, DKFZp434J1313, FLIP1, KLIP, MGC4289ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-288-409