Learn More
Aviva Systems Biology Corp TNKS Antibody - middle region (ARP33755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNKS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNKS. Alternate names: PARP-5a, PARP5A, PARPL, TIN1, TINF1, TNKS1, pART5ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55139