Learn More
Sigma Aldrich Fine Chemicals Biosciences TNM-fH insect medium With
Catalog No. 502901776
Encompass
A number of modifications of Graces medium were developed in several laboratories in the late 1960s and 1970s. Many of these modifications were developed by Dr. W.f. Hink for the growth of cells derived from the cabbage looper Tricoplusia ni. This formula represents the modification known as TNM-fH. The medium when properly supplemented has been found to support the growth of cells derived from a variety of lepidopteran species.
Catalog No. 50-290-1776 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T103210L
A number of modifications of Graces medium were developed in several laboratories in the late 1960s and 1970s. Many of these modifications were developed by Dr. W.f. Hink for the growth of cells derived from the cabbage looper Tricoplusia ni. This formula represents the modification known as TNM-fH. The medium when properly supplemented has been found to support the growth of cells derived from a variety of lepidopteran species.