Learn More
Aviva Systems Biology Corp TNMD Antibody - middle region (ARP49697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNMD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNMD. Alternate names: BRICD4, CHM1-LIKE, CHM1L, TEMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60146