Learn More
Biomatik Corporation Tnmd Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11138
Encompass_Preferred
Tnmd Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TNMD. The Tnmd Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tenomodulin Protein (131-254Aa). Applications: ELISA, WB, IF. Target Synonyms: TNMD. Accession Number: Q9H2S6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9251