Learn More
Aviva Systems Biology Corp TNN Antibody - N-terminal region (ARP63200_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63200_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNN. It was validated on Western Blot. . Alternate names: TN-WApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-287-909