Learn More
Aviva Systems Biology Corp TNNI3 Antibody - middle region (ARP41391_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41391_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNNI3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNNI3. Alternate names: CMH7, MGC116817, TNNC1, cTnI, RCM1, CMD2A, CMD1FFApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57829