Learn More
Aviva Systems Biology Corp TNNI3K Antibody - N-terminal region (ARP48044_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48044_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNNI3K. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TNNI3K. Alternate names: CARK, MGC142099, MGC33828ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-341