Learn More
Aviva Systems Biology Corp TNNT1 Antibody - middle region (ARP42120_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42120_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNNT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TNNT1. Alternate names: ANM, MGC104241, TNT, STNT, TNTSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-58114