Learn More
Aviva Systems Biology Corp TNNT2 Antibody - N-terminal region (ARP41717_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41717_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNNT2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNNT2. Alternate names: CMH2, CMPD2, MGC3889, TnTC, cTnT, RCM3, LVNC6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57965