Learn More
Aviva Systems Biology Corp TNNT3 Antibody - C-terminal region (ARP51286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNNT3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TNNT3. Alternate names: AMCD2B, DA2B, DKFZp779M2348, FSSV, TNTFApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60535