Learn More
Cayman Chemical tnofovIr DIsoproxIlfumart 10mg
SDP
Catalog No. 502966426
Encompass_Preferred
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes
Catalog No. 50-296-6426 Supplier Cayman Chemical Supplier No. 1692210 MG
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes