Learn More
Cayman Chemical Tenofovir Disoproxil (fumarate), Purity: ≥98%, CAS: 202138-50-9, 50 mg
SDP
Catalog No. 502967443
Encompass_Preferred
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes
Catalog No. 50-296-7443 Supplier Cayman Chemical Supplier No. 1692250 MG
A prodrug of the HIV reverse transcriptase and hepatitis B virus polymerase inhibitor, tenofovir; >100-fold greater anti-HIV activity than that of poorly bioavailable tenofovir in a T cell line and primary blood lymphocytes