Learn More
Medchemexpress LLC HY-101932 10mg Medchemexpress, TNP-470 CAS:129298-91-5 Purity:>98%
SDP
Catalog No. 501967925
Encompass_Preferred
Medchemexpress, HY-101932 10mg TNP-470 CAS:129298-91-5 TNP-470 is a methionine aminopeptidase-2 inhibitor and also an angiogenesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7925 Supplier Medchemexpress LLC Supplier No. HY10193210MG
Add to Cart
Add to Cart
Medchemexpress, HY-101932 10mg TNP-470 CAS:129298-91-5 TNP-470 is a methionine aminopeptidase-2 inhibitor and also an angiogenesis inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.