Learn More
Medchemexpress LLC HY-110079 5mg Medchemexpress, TNP CAS:519178-28-0 Purity:>98%
SDP
Catalog No. 501968741
Encompass_Preferred
Medchemexpress, HY-110079 5mg TNP CAS:519178-28-0 TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 µM and 10.2 µM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8741 Supplier Medchemexpress LLC Supplier No. HY1100795MG
Medchemexpress, HY-110079 5mg TNP CAS:519178-28-0 TNP is a cell-permeable inhibitor of IP6K1 and IP3K, with IC50 values of 0.55 µM and 10.2 µM for IP3K, respectively. TNP binds to the ATP-binding sites of both enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.