Learn More
Cayman Chemical TNP-GTPsodIum salt 700ug
Supplier: Cayman Chemical 38570700 UG
A fluorescent derivative of GTP; an antagonist of the purinergic P2X2 and P2X2/3 receptors (IC50 = 0.4 and 1.2 nM, respectively); selectively inhibits rat sGC (Ki = 11 nM) over bovine liver GDH (Ki = 2.7 µM) and the calmodulin-dependent B. pertussis AC toxin (Kis = 20 and 320 µM in the presence of manganese or magnesium, respectively; ex/em = 470/552 nm, respectively