Learn More
ABclonal Technology TNR Rabbit pAb
SDP
Supplier:  ABclonal Technology A15324500UL
Encompass_Preferred
This gene encodes a member of the tenascin family of extracellular matrix glycoproteins. The encoded protein is restricted to the central nervous system. The protein may play a role in neurite outgrowth, neural cell adhesion and modulation of sodium channel function. It is a constituent of perineuronal nets.
Catalog No. 50-001-08005