Learn More
Aviva Systems Biology Corp TNRC5 Antibody - C-terminal region (ARP34422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNRC5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TNRC5. Alternate names: CAG4A, ERDA5, TNRC5, PRAT4AApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-52955