Learn More
Aviva Systems Biology Corp TNRC6A Antibody - N-terminal region (ARP40956_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40956_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNRC6A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNRC6A. Alternate names: CAGH26, DKFZp666E117, FLJ22043, GW1, GW182, KIAA1460, MGC75384, TNRC6ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse
Catalog No. 50-014-53254