Learn More
Aviva Systems Biology Corp TNRC6B Antibody - N-terminal region (ARP40804_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40804_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNRC6B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TNRC6B. Alternate names: KIAA1093ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53238