Learn More
Promocell GmbH PromoCell TNS 125 ml, Trypsin Neutralizing Solution (0.05 % trypsin inhibitor, 0.1 % BSA)
Catalog No. 50306384
Encompass
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.Trypsin Neutralizing Solution containing 0.05% Trypsin Inhibitor in 0.1% BSA.
Catalog No. 50-306-384 Supplier Promocell GmbH Supplier No. C41110
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.Trypsin Neutralizing Solution containing 0.05% Trypsin Inhibitor in 0.1% BSA.