Learn More
Aviva Systems Biology Corp TNS4 Antibody - N-terminal region (ARP60705_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60705_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TNS4. It was validated on Western Blot. . Alternate names: CTEN, FLJ14950, PP14434ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-287-134