Learn More
BIOLOGIX USA INC 1ml Individually Wrapped Transfer Pipets, Sterile, 2000/case
Catalog No. NC3762218
Encompass_Preferred
1ml Individually Wrapped Transfer PipetsMaterial: PE, will not shatter or breakAn excellent alternative to Pasteur pipettes and rubber bulbsEvenly distributed thickness ensures uniform suction forceGamma radiation sterilized Wrapped individually (sterile) DNase & RNase freeTransparent wall permits easy viewing of pipette contentsWith molded graduations on the body of the transfer pipettes
Catalog No. NC3762218 Supplier BIOLOGIX USA INC Supplier No. 300135A1
1ml Individually Wrapped Transfer PipetsMaterial: PE, will not shatter or breakAn excellent alternative to Pasteur pipettes and rubber bulbsEvenly distributed thickness ensures uniform suction forceGamma radiation sterilized Wrapped individually (sterile) DNase & RNase freeTransparent wall permits easy viewing of pipette contentsWith molded graduations on the body of the transfer pipettes