Learn More
Cayman Chemical TnshInon IIA 10mg
SDP
Catalog No. 502955442
Encompass_Preferred
A natural product with anti-inflammatory and cytotoxic activity; inhibits production of TNFα, IL-1β, IL-6, NO, INFγ, and expression of iNOS, and IL-12; blocks human aortic smooth muscle cell migration, inhibits MMP-9 activity, and interferes with PI3K/Akt and ERK1/2 signaling pathways
Catalog No. 50-295-5442 Supplier Cayman Chemical Supplier No. 1432310 MG
A natural product with anti-inflammatory and cytotoxic activity; inhibits production of TNFα, IL-1β, IL-6, NO, INFγ, and expression of iNOS, and IL-12; blocks human aortic smooth muscle cell migration, inhibits MMP-9 activity, and interferes with PI3K/Akt and ERK1/2 signaling pathways