Learn More
Biotang Inc TNT buffer, 1000 ML,BM-309T, 10 mM Tris-HCl, 150 mM NaCl, 0.05% Tween-20. pH 8.0 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50155874 Shop All BIOTANG Inc Products
Encompass_Preferred
TNT buffer, 1000 ML,BM-309T, 10 mM Tris-HCl, 150 mM NaCl, 0.05% Tween-20. pH 8.0 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.
Catalog No. 50-155-874 Supplier Biotang Inc Supplier No. BTBB552
TNT buffer, 1000 ML,BM-309T, 10 mM Tris-HCl, 150 mM NaCl, 0.05% Tween-20. pH 8.0 +/- 0.2. Prepared in 18.2 megohms water and filtered through 1 micron filter. Shipped at room temperature. Store at room temperature.