Learn More
Hach Company Phosphorus (Reactive) TNT Reagent Set, High Range Molybdovanadate
Catalog No. 501996647 Shop All Hach Company Products
Encompass
For determination of Reactive Phosphorus (ortho-phosphate) by the Molybdovanadate Test 'N Tube™ Method. Hach Method 8114. Range: 1.0 - 100.0 mg/L PO4. Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).
Catalog No. 50-199-6647 Supplier Hach Company Supplier No. 2767345
For determination of Reactive Phosphorus (ortho-phosphate) by the Molybdovanadate Test 'N Tube™ Method. Hach Method 8114. Range: 1.0 - 100.0 mg/L PO4. Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).