Learn More
Hach Company Phosphorus (Total) TNT Reagent Set, High Range Molybdovanadate
Supplier:  Hach Company 2767245
Encompass
For determination of Total Phosphorus (phosphate) by the Molybdovanadate Test 'N Tube™ Method. Hach Method 10127. Range: 1.0 - 100.0 mg/L PO4 Set includes vials and reagents for 25-50 tests (dependent upon the number of reagent blank tests performed).
Catalog No. 50-199-6646