Learn More
Hach Company Metals Prep Set for TNTplus Vial Test, 50 Digestions
Supplier:  Hach Company TNT890
Encompass
Sample preparation set for the determination of total heavy metals with TNTplus metal tests: TNT850 Lead, TNT852 Cadmium, TNT858 Iron, TNT860 Copper, TNT856 Nickel. For 50 digestions.
Catalog No. 50-199-8167