Learn More
Hach Company Phosphorus (Total and Acid Hydrolyzable) TNT Reagent Set, Low Range Ascorbic Acid Method
Catalog No. 501996602 Shop All Hach Company Products
Encompass
For determination of Total and Acid-Hydrolyzable Phosphorus (Phosphate) by the PhosVer 3 Ascorbic Acid method with Acid Hydrolysis and Persulfate Digestion. Hach Method 8180. Range: 0.06 - 3.50 mg/L PO4. Test 'N Tube™ Reagent Set contains reagents for sample digestion and photometric analyses. Up to 50 tests.
Catalog No. 50-199-6602 Supplier Hach Company Supplier No. 2742745
For determination of Total and Acid-Hydrolyzable Phosphorus (Phosphate) by the PhosVer 3 Ascorbic Acid method with Acid Hydrolysis and Persulfate Digestion. Hach Method 8180. Range: 0.06 - 3.50 mg/L PO4. Test 'N Tube™ Reagent Set contains reagents for sample digestion and photometric analyses. Up to 50 tests.