Learn More
Hach Company Phosphorus (Reactive) TNT Reagent Set, Low Range
Supplier:  Hach Company 2742545
Encompass
For determination of Reactive Phosphorus (ortho-phosphate) by the PhosVer 3 Ascorbic Acid Test 'N Tube™ method. Hach Method 8048. Range: 0.06 - 5.00 mg/L PO4. Set includes: PhosVer 3 Phosphate Reagent Powder Pillows and Reactive Phosphorus Test 'N Tube Dilution Vials for up to 50 tests.
Catalog No. 50-199-6600