Learn More
Hach Company Phosphorus (Total) TNT Reagent Set, Low Range
Supplier:  Hach Company 2742645
Encompass
For determination of Total Phosphorus by the PhosVer 3 Ascorbic Acid method with Acid Persulfate Digestion. EPA CompliantHach Method 8190. Range: 0.06 - 3.50 mg/L PO4.Test 'N Tube™ Reagent Set contains reagents for sample digestion and photometric analyses. Up to 50 tests.
Catalog No. 50-199-6601