Learn More
Hach Company Fluoride TNTplus Vial Test (0.1-2.5 mg/L F), 25 Tests
Supplier:  Hach Company TNT878
Encompass
Fluoride TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Fluoride by the SPANDS 2 method. The SPADNS 2 reagent formulation removes interference from chlorine without arsenic. EPA Compliant for use in Drinking Water (40 CFR 141) and Wastewater reporting programs (SM 4500-F B or C).Reduce human errors and contamination from competing methods requiring the use of multiple pipettes or separate ampules for blanks and samples!
Catalog No. 50-199-8162