Learn More
Medchemexpress LLC TO-1187 (TFA) | 98.4% | 795.72 | 25 MG
SDP
Catalog No. 5000384321
Encompass_Preferred
TO-1187 TFA is a selective PROTAC degrader designed to target HDAC6, effectively promoting its ubiquitination and subsequent degradation. It is a valuable tool for research into hematological malignancies and solid tumors, composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective PROTAC degrader for HDAC6
  • Promotes ubiquitination and degradation of HDAC6
  • Useful for studying hematological malignancies and solid tumors
  • Provided as a light yellow to yellow solid
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-003-84321 Supplier Medchemexpress LLC Supplier No. 25MGHY173266A
TO-1187 TFA is a selective PROTAC degrader designed to target HDAC6, effectively promoting its ubiquitination and subsequent degradation. It is a valuable tool for research into hematological malignancies and solid tumors, composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective PROTAC degrader for HDAC6
  • Promotes ubiquitination and degradation of HDAC6
  • Useful for studying hematological malignancies and solid tumors
  • Provided as a light yellow to yellow solid
  • Soluble in DMSO at 100 mg/mL