Learn More
Medchemexpress LLC To-1187 (TFA) | 98.4% | 795.72 | 50 MG
SDP
Catalog No. 5000385378
Encompass_Preferred
TO-1187 TFA is a selective HDAC6 PROTAC degrader with a DC50 of 5.81 nM. It functions by promoting the ubiquitination and degradation of HDAC6. This compound is suitable for use in research studying hematological malignancies and solid tumors. The compound is composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective HDAC6 PROTAC degrader, exhibits a DC50 of 5.81 nM, indicating potent and specific degradation of HDAC6.
  • Promotes ubiquitination and subsequent degradation of HDAC6.
  • Useful in the study of hematological malignancies and solid tumors.
  • Consists of a specific HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Purity of 98.41%.
Catalog No. 50-003-85378 Supplier Medchemexpress LLC Supplier No. HY173266A50MG
TO-1187 TFA is a selective HDAC6 PROTAC degrader with a DC50 of 5.81 nM. It functions by promoting the ubiquitination and degradation of HDAC6. This compound is suitable for use in research studying hematological malignancies and solid tumors. The compound is composed of an HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Selective HDAC6 PROTAC degrader, exhibits a DC50 of 5.81 nM, indicating potent and specific degradation of HDAC6.
  • Promotes ubiquitination and subsequent degradation of HDAC6.
  • Useful in the study of hematological malignancies and solid tumors.
  • Consists of a specific HDAC6 ligand, a CRBN ligase ligand, and a linker.
  • Purity of 98.41%.