Learn More
Medchemexpress LLC TO-1187 (TFA) | 98.4% | 795.72 | C36H36F3N9O9 | 10 MG
SDP
Catalog No. 5000295449
Encompass_Preferred
TO-1187 TFA is a selective PROTAC degrader that targets HDAC6 and recruits cereblon to promote ubiquitination and proteasomal degradation of HDAC6. Supplied as a trifluoroacetic acid salt in solid form, it is intended as a research tool for cellular and preclinical studies investigating HDAC6 biology and PROTAC-mediated target removal.
  • Selective HDAC6 degrader with DC50 of 5.81 nM.
  • Engages cereblon to induce ubiquitination and proteasomal degradation.
  • High chemical purity (98.4%) suitable for biochemical and cellular assays.
  • Provided as a solid TFA salt for stable handling and storage.
  • Includes analytical documentation such as COA, H NMR, LCMS, and RP-HPLC.
  • Storage recommended at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-95449 Supplier Medchemexpress LLC Supplier No. HY173266A10MG
TO-1187 TFA is a selective PROTAC degrader that targets HDAC6 and recruits cereblon to promote ubiquitination and proteasomal degradation of HDAC6. Supplied as a trifluoroacetic acid salt in solid form, it is intended as a research tool for cellular and preclinical studies investigating HDAC6 biology and PROTAC-mediated target removal.
  • Selective HDAC6 degrader with DC50 of 5.81 nM.
  • Engages cereblon to induce ubiquitination and proteasomal degradation.
  • High chemical purity (98.4%) suitable for biochemical and cellular assays.
  • Provided as a solid TFA salt for stable handling and storage.
  • Includes analytical documentation such as COA, H NMR, LCMS, and RP-HPLC.
  • Storage recommended at 4°C under nitrogen; in solvent store at -80°C (6 months) or -20°C (1 month).