Learn More
Medchemexpress LLC TO-PRO-3 iodide 671ug | 157199-63-8 | 671.42 g·mol⁻¹ | C26H31I2N3S | 671 UG
SDP
Catalog No. 5000229411
Encompass_Preferred
TO-PRO-3 iodide is a far-red monomeric cyanine nucleic acid stain that selectively binds nucleic acids and produces bright nuclear fluorescence in fixed or membrane-compromised cells.
  • Provides bright far-red nuclear staining with excitation ≈642 nm and emission ≈661 nm.
  • Is cell-impermeant and useful as a viability marker and nuclear counterstain.
  • Is suitable for flow cytometry, fluorescence microscopy, and chromosome staining.
  • Is supplied as a DMSO solution for convenient dilution to working concentrations.
  • Offers high fluorescence enhancement upon nucleic acid binding for sensitive detection.
  • Is provided in a small vial format (e.g., 671 μg, 5 mM x 200 μL) for research use.
Catalog No. 50-002-29411 Supplier Medchemexpress LLC Supplier No. HY117070671UG
TO-PRO-3 iodide is a far-red monomeric cyanine nucleic acid stain that selectively binds nucleic acids and produces bright nuclear fluorescence in fixed or membrane-compromised cells.
  • Provides bright far-red nuclear staining with excitation ≈642 nm and emission ≈661 nm.
  • Is cell-impermeant and useful as a viability marker and nuclear counterstain.
  • Is suitable for flow cytometry, fluorescence microscopy, and chromosome staining.
  • Is supplied as a DMSO solution for convenient dilution to working concentrations.
  • Offers high fluorescence enhancement upon nucleic acid binding for sensitive detection.
  • Is provided in a small vial format (e.g., 671 μg, 5 mM x 200 μL) for research use.