Learn More
Aviva Systems Biology Corp TOB1 Antibody - middle region (ARP52105_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52105_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TOB1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TOB1. Alternate names: APRO6, MGC104792, MGC34446, PIG49, TOB, TROB, TROB1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-285-636